1-Boc-2-piperidone 98%

BD40415-100G BD40415-500G
BLDPBD40415-100GEA 323 EUR
BLDPBD40415-100G BLDPBD40415-500G
1-Boc-2-piperidone 98%
1-Boc-2-piperidone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD02179046
CAS Number: 85908-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR