2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine 98%

BD4062-100G BD4062-1G
BLDPBD4062-100GEA 234 EUR
BLDPBD4062-100G BLDPBD4062-1G
2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine 98%
2-[N,N-Bis(trifluoromethanesulphonyl)amino]-5-chloropyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₃ClF₆N₂O₄S₂
MW: 392.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191833
CAS Number: 145100-51-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR