Benzhydrylamine hydrochloride 97%
|
|
BD40651-10G
BD40651-5G
BD40651-100G
BLDPBD40651-10GEA
7.4
EUR
BLDPBD40651-10G
BLDPBD40651-5G
BLDPBD40651-100G
Benzhydrylamine hydrochloride 97%
Benzhydrylamine hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₁₃H₁₄NCl Storage Temperature: Ambient |
MDL Number:
MFCD00013023 CAS Number: 5267-34-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...