3(2H)-Pyridazinone 97%

BD41033-1G BD41033-100G
BLDPBD41033-1GEA 7.4 EUR
BLDPBD41033-1G BLDPBD41033-100G
3(2H)-Pyridazinone 97%
3(2H)-Pyridazinone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₄H₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD00099605
CAS Number: 504-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR