1-(6-Methylpyridin-3-yl)-2-(4-(methylsulphonyl)phenyl)ethanone 97%
|
|
BD41468-25G
BD41468-1G
BLDPBD41468-25GEA
214
EUR
BLDPBD41468-25G
BLDPBD41468-1G
1-(6-Methylpyridin-3-yl)-2-(4-(methylsulphonyl)phenyl)ethanone 97%
1-(6-Methylpyridin-3-yl)-2-(4-(methylsulphonyl)phenyl)ethanone
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₅H₁₅NO₃S Storage Temperature: Ambient |
MDL Number:
MFCD11110696 CAS Number: 221615-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...