2-Bromo-m-xylene 98%
|
|
BD41810-10G
BD41810-500G
BLDPBD41810-10GEA
8.4
EUR
BLDPBD41810-10G
BLDPBD41810-500G
2-Bromo-m-xylene 98%
2-Bromo-m-xylene
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₉Br Storage Temperature: Ambient |
MDL Number:
MFCD00000075 CAS Number: 576-22-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...