2-Amino-3,5-difluoropyridine 98%

BD41977-5G BD41977-10G
BLDPBD41977-5GEA 81.9 EUR
BLDPBD41977-5G BLDPBD41977-10G
2-Amino-3,5-difluoropyridine 98%
2-Amino-3,5-difluoropyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₅H₄F₂N₂
MW: 130.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788760
CAS Number: 732306-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR