N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine 97%
|
|
BD42364-100MG
BD42364-250MG
BLDPBD42364-100MGEA
75.6
EUR
BLDPBD42364-100MG
BLDPBD42364-250MG
N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine 97%
N-Boc-(S)-2,4,5-trifluoro-b-homophenylalanine
Storage condition: Sealed in dry, 2-8°C
Formula:
C₁₅H₁₈F₃NO₄ MW: 333.303 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06659150 CAS Number: 922178-94-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...