5-Chloro-6-(chloromethyl)pyrimidine-2,4(1H,3H)-dione 95%

BD427244-5G BD427244-25G
BLDPBD427244-5GEA 55.7 EUR
BLDPBD427244-5G BLDPBD427244-25G
5-Chloro-6-(chloromethyl)pyrimidine-2,4(1H,3H)-dione 95%
5-Chloro-6-(chloromethyl)pyrimidine-2,4(1H,3H)-dione

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₄Cl₂N₂O₂
MDL Number: MFCD01571383
CAS Number: 73742-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR