4-Bromosalicylaldehyde 98%

BD4319-500G BD4319-1G
BLDPBD4319-500GEA 1610 EUR
BLDPBD4319-500G BLDPBD4319-1G
4-Bromosalicylaldehyde 98%
4-Bromosalicylaldehyde

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₅BrO₂
Storage Temperature: Ambient
MDL Number: MFCD06252606
CAS Number: 22532-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR