3,4-Dichlorobenzamide 98+%

BD4333-5G BD4333-250MG
BLDPBD4333-5GEA 126 EUR
BLDPBD4333-5G BLDPBD4333-250MG
3,4-Dichlorobenzamide 98+%
3,4-Dichlorobenzamide

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₅Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD00053009
CAS Number: 2670-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR