(S)-2-Amino-3-(1H-pyrrolo[2,3-c]pyridin-3-yl)propanoic acid 95%

BD435022-50MG BD435022-25MG
BLDPBD435022-50MGEA 392 EUR
BLDPBD435022-50MG BLDPBD435022-25MG
(S)-2-Amino-3-(1H-pyrrolo[2,3-c]pyridin-3-yl)propanoic acid 95%
(S)-2-Amino-3-(1H-pyrrolo[2,3-c]pyridin-3-yl)propanoic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₁N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD29071118
CAS Number: 149704-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR