4-Amino-2-chlorobenzonitrile 98%

BD4368-500G BD4368-5G
BLDPBD4368-500GEA 816 EUR
BLDPBD4368-500G BLDPBD4368-5G
4-Amino-2-chlorobenzonitrile 98%
4-Amino-2-chlorobenzonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₅ClN₂
Storage Temperature: Ambient
MDL Number: MFCD00035926
CAS Number: 20925-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR