2-Amino-4-chlorobenzothiazole 97%

BD4379-100G BD4379-25G
BLDPBD4379-100GEA 167 EUR
BLDPBD4379-100G BLDPBD4379-25G
2-Amino-4-chlorobenzothiazole 97%
2-Amino-4-chlorobenzothiazole

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₅ClN₂S
Storage Temperature: Ambient
MDL Number: MFCD00005791
CAS Number: 19952-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR