(2-Methyl-1,3-thiazol-4-yl)methanol 96%

BD44415-1G BD44415-5G
BLDPBD44415-1GEA 69.3 EUR
BLDPBD44415-1G BLDPBD44415-5G
(2-Methyl-1,3-thiazol-4-yl)methanol 96%
(2-Methyl-1,3-thiazol-4-yl)methanol

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₇NOS
Storage Temperature: Refrigerator
MDL Number: MFCD08703637
CAS Number: 76632-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR