Di-tert-butyl succinate 95%

BD446885-1G BD446885-5G
BLDPBD446885-1GEA 179 EUR
BLDPBD446885-1G BLDPBD446885-5G
Di-tert-butyl succinate 95%
Di-tert-butyl succinate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₂₂O₄
MW: 230.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00127979
CAS Number: 926-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR