10-Bromo-5H-dibenzo[b,f]azepine-5-carboxamide 97%
BD449992-1G
BD449992-250MG
BLDPBD449992-1GEA
519
EUR
BLDPBD449992-1G
BLDPBD449992-250MG
10-Bromo-5H-dibenzo[b,f]azepine-5-carboxamide 97%
10-Bromo-5H-dibenzo[b,f]azepine-5-carboxamide
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₅H₁₁BrN₂O |
MDL Number:
MFCD12032130 CAS Number: 59690-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...