(3-Bromopropyl)triphenylphosphonium bromide 97%

BD45062-1000G BD45062-500G
BLDPBD45062-1000GEA 406 EUR
BLDPBD45062-1000G BLDPBD45062-500G
(3-Bromopropyl)triphenylphosphonium bromide 97%
(3-Bromopropyl)triphenylphosphonium bromide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₁H₂₁Br₂P
MW: 464.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011866
CAS Number: 3607-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR