3-Bromoanthranilic acid 97%

BD4524-1G BD4524-500G
BLDPBD4524-1GEA 7.4 EUR
BLDPBD4524-1G BLDPBD4524-500G
3-Bromoanthranilic acid 97%
3-Bromoanthranilic acid

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₇H₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD03618453
CAS Number: 20776-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR