2-Bromo-5-nitroanisole 98%

BD4535-1G BD4535-10G
BLDPBD4535-1GEA 6.3 EUR
BLDPBD4535-1G BLDPBD4535-10G
2-Bromo-5-nitroanisole 98%
2-Bromo-5-nitroanisole

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00041250
CAS Number: 77337-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR