3-Chloro-2-iodotoluene 99%
|
|
BD4574-10G
BD4574-500G
BLDPBD4574-10GEA
52.5
EUR
BLDPBD4574-10G
BLDPBD4574-500G
3-Chloro-2-iodotoluene 99%
3-Chloro-2-iodotoluene
Storage condition: Keep in dark place, Sealed in dry, 2-8°C
Formula:
C₇H₆ClI Storage Temperature: Refrigerator |
MDL Number:
MFCD00051756 CAS Number: 5100-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...