4-Chloroanthranilic acid 95%
|
|
BD4590-100G
BD4590-25G
BLDPBD4590-100GEA
24.2
EUR
BLDPBD4590-100G
BLDPBD4590-25G
4-Chloroanthranilic acid 95%
4-Chloroanthranilic acid
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₇H₆ClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD00007778 CAS Number: 89-77-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...