2-Cyano-3-methylpyridine 98%

BD4662-100G BD4662-10G
BLDPBD4662-100GEA 28.4 EUR
BLDPBD4662-100G BLDPBD4662-10G
2-Cyano-3-methylpyridine 98%
2-Cyano-3-methylpyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₆N₂
Storage Temperature: Ambient
MDL Number: MFCD00190585
CAS Number: 20970-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR