(Ethoxycarbonylmethyl)triphenylphosphonium bromide 98%

BD46690-5G BD46690-500G
BLDPBD46690-5GEA 7.4 EUR
BLDPBD46690-5G BLDPBD46690-500G
(Ethoxycarbonylmethyl)triphenylphosphonium bromide 98%
(Ethoxycarbonylmethyl)triphenylphosphonium bromide

Storage condition: Inert atmosphere, Room Temperature

Formula: C₂₂H₂₂BrO₂P
Storage Temperature: Ambient
MDL Number: MFCD00011835
CAS Number: 1530-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR