Methyl 3-chloro-4-methylthiophene-2-carboxylate 98%

BD4784-25G BD4784-10G
BLDPBD4784-25GEA 287 EUR
BLDPBD4784-25G BLDPBD4784-10G
Methyl 3-chloro-4-methylthiophene-2-carboxylate 98%
Methyl 3-chloro-4-methylthiophene-2-carboxylate

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₇ClO₂S
Storage Temperature: Ambient
MDL Number: MFCD00052072
CAS Number: 175137-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR