4-Fluorobenzhydrazide 97%

BD4798-10G BD4798-100G
BLDPBD4798-10GEA 26.3 EUR
BLDPBD4798-10G BLDPBD4798-100G
4-Fluorobenzhydrazide 97%
4-Fluorobenzhydrazide

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₇H₇FN₂O
Storage Temperature: Ambient
MDL Number: MFCD00060562
CAS Number: 456-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR