(S)-(-)-1-(2-Naphthyl)ethylamine 97%

BD47985-25G BD47985-5G
BLDPBD47985-25GEA 286 EUR
BLDPBD47985-25G BLDPBD47985-5G
(S)-(-)-1-(2-Naphthyl)ethylamine 97%
(S)-(-)-1-(2-Naphthyl)ethylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₁₂H₁₃N
Storage Temperature: Ambient
MDL Number: MFCD00085366
CAS Number: 3082-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR