(S)-tert-Butyl (1,5-dihydroxypentan-2-yl)carbamate 98%

BD48032-1G BD48032-250MG
BLDPBD48032-1GEA 83 EUR
BLDPBD48032-1G BLDPBD48032-250MG
(S)-tert-Butyl (1,5-dihydroxypentan-2-yl)carbamate 98%
(S)-tert-Butyl (1,5-dihydroxypentan-2-yl)carbamate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD03093945
CAS Number: 162955-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR