(S)-(-)-4-Bromo-α-methylbenzyl alcohol 98%

BD48383-5G BD48383-25G
BLDPBD48383-5GEA 185 EUR
BLDPBD48383-5G BLDPBD48383-25G
(S)-(-)-4-Bromo-α-methylbenzyl alcohol 98%
(S)-(-)-4-Bromo-α-methylbenzyl alcohol

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₉BrO
Storage Temperature: Ambient
MDL Number: MFCD06201860
CAS Number: 100760-04-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR