(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride 97%

BD4909-100MG BD4909-10G
BLDPBD4909-100MGEA 69.3 EUR
BLDPBD4909-100MG BLDPBD4909-10G
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride 97%
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₇H₉BClNO₂
MW: 185.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04115645
CAS Number: 117098-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR