N-Methoxycarbonyl-L-tert-leucine 97%
|
|
BD49202-100G
BD49202-5G
BLDPBD49202-100GEA
119
EUR
BLDPBD49202-100G
BLDPBD49202-5G
N-Methoxycarbonyl-L-tert-leucine 97%
N-Methoxycarbonyl-L-tert-leucine
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₈H₁₅NO₄ MW: 189.211 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07778455 CAS Number: 162537-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...