(4-Nitro-1H-pyrazol-1-yl)acetonitrile 97%

BD494609-1G BD494609-25G
BLDPBD494609-1GEA 111 EUR
BLDPBD494609-1G BLDPBD494609-25G
(4-Nitro-1H-pyrazol-1-yl)acetonitrile 97%
(4-Nitro-1H-pyrazol-1-yl)acetonitrile

Storage condition: Sealed in dry, 2-8°C

Formula: C₅H₄N₄O₂
Storage Temperature: Refrigerator
MDL Number: MFCD04969696
CAS Number: 1001500-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR