4-(2,6-Dichlorophenyl)thiazol-2-amine 97%

BD495404-1G BD495404-100MG
BLDPBD495404-1GEA 569 EUR
BLDPBD495404-1G BLDPBD495404-100MG
4-(2,6-Dichlorophenyl)thiazol-2-amine 97%
4-(2,6-Dichlorophenyl)thiazol-2-amine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₉H₆Cl₂N₂S
Storage Temperature: Ambient
MDL Number: MFCD06739834
CAS Number: 736901-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR