5-Methoxy-2-nitrophenylamine 97%

BD4983-25G BD4983-1G BD4983-100G
BLDPBD4983-25GEA 75.6 EUR
BLDPBD4983-25G BLDPBD4983-1G BLDPBD4983-100G
5-Methoxy-2-nitrophenylamine 97%
5-Methoxy-2-nitrophenylamine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C7H8N2O3
MW: 168.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00179573
CAS Number: 16133-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR