4'-Amino-2'-hydroxyacetophenone 96%

BD50785-25G BD50785-1G
BLDPBD50785-25GEA 404 EUR
BLDPBD50785-25G BLDPBD50785-1G
4'-Amino-2'-hydroxyacetophenone 96%
4'-Amino-2'-hydroxyacetophenone

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₈H₉NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00100636
CAS Number: 2476-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR