1-(tert-Butoxycarbonyl)-2-pyrrolidinone 95%

BD51860-100G BD51860-500G
BLDPBD51860-100GEA 44.1 EUR
BLDPBD51860-100G BLDPBD51860-500G
1-(tert-Butoxycarbonyl)-2-pyrrolidinone 95%
1-(tert-Butoxycarbonyl)-2-pyrrolidinone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₉H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00674099
CAS Number: 85909-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR