1-Boc-indole-3-boronic acid 97%

BD51870-250MG BD51870-100MG
BLDPBD51870-250MGEA 22.1 EUR
BLDPBD51870-250MG BLDPBD51870-100MG
1-Boc-indole-3-boronic acid 97%
1-Boc-indole-3-boronic acid

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₁₃H₁₆BNO₄
Storage Temperature: Freezer
MDL Number: MFCD06656257
CAS Number: 181365-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR