p-Anisohydrazide 98%

BD5189-25G BD5189-500G
BLDPBD5189-25GEA 20 EUR
BLDPBD5189-25G BLDPBD5189-500G
p-Anisohydrazide 98%
p-Anisohydrazide

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00017073
CAS Number: 3290-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR