1H-1,2,3-Triazole-4,5-dicarboxylic acid 97%

BD52798-100MG BD52798-1G
BLDPBD52798-100MGEA 39.9 EUR
BLDPBD52798-100MG BLDPBD52798-1G
1H-1,2,3-Triazole-4,5-dicarboxylic acid 97%
1H-1,2,3-Triazole-4,5-dicarboxylic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₄H₃N₃O₄
Storage Temperature: Refrigerator
MDL Number: MFCD00005246
CAS Number: 4546-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR