1,2,3-Trifluorobenzene 98%

BD52813-25G BD52813-10G
BLDPBD52813-25GEA 43.1 EUR
BLDPBD52813-25G BLDPBD52813-10G
1,2,3-Trifluorobenzene 98%
1,2,3-Trifluorobenzene

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₃F₃
Storage Temperature: Refrigerator
MDL Number: MFCD00012013
CAS Number: 1489-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR