4-Methoxy-2-methylpyrimidine-5-carbonitrile 95%

BD528634-250MG BD528634-100MG
BLDPBD528634-250MGEA 39.9 EUR
BLDPBD528634-250MG BLDPBD528634-100MG
4-Methoxy-2-methylpyrimidine-5-carbonitrile 95%
4-Methoxy-2-methylpyrimidine-5-carbonitrile

Storage condition: Sealed in dry, 2-8°C

Formula: C₇H₇N₃O
Storage Temperature: Refrigerator
MDL Number: MFCD18377705
CAS Number: 2006-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR