1,3,4,6-Tetra-O-acetyl-β-D-mannopyranose 97%

BD53560-1G BD53560-250MG
BLDPBD53560-1GEA 93.5 EUR
BLDPBD53560-1G BLDPBD53560-250MG
1,3,4,6-Tetra-O-acetyl-β-D-mannopyranose 97%
1,3,4,6-Tetra-O-acetyl-beta-D-mannopyranose

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₄H₂₀O₁₀
Storage Temperature: Refrigerator
MDL Number: MFCD00012354
CAS Number: 18968-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR