Benzo[d][1,3]dioxol-4-amine 97%

BD53743-1G BD53743-100MG
BLDPBD53743-1GEA 152 EUR
BLDPBD53743-1G BLDPBD53743-100MG
Benzo[d][1,3]dioxol-4-amine 97%
Benzo[d][1,3]dioxol-4-amine

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₇H₇NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09038121
CAS Number: 1668-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR