1,3-Benzodioxole 98%

BD53746-500G BD53746-1000G
BLDPBD53746-500GEA 49.4 EUR
BLDPBD53746-500G BLDPBD53746-1000G
1,3-Benzodioxole 98%
1,3-Benzodioxole

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₆O₂
Storage Temperature: Ambient
MDL Number: MFCD00005818
CAS Number: 274-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR