1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride 95%

BD53781-1G BD53781-10G
BLDPBD53781-1GEA 9.5 EUR
BLDPBD53781-1G BLDPBD53781-10G
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride 95%
1,3-Bis(2,6-diisopropylphenyl)imidazolinium chloride

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₂₇H₃₉ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD07369796
CAS Number: 258278-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR