1,3-Phenylenediacetic acid 97%

BD54290-10G BD54290-1G
BLDPBD54290-10GEA 74.6 EUR
BLDPBD54290-10G BLDPBD54290-1G
1,3-Phenylenediacetic acid 97%
1,3-Phenylenediacetic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₀H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD00004341
CAS Number: 19806-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR