2-Chloro-1,4-dibromobenzene 98%
|
|
BD54595-100G
BD54595-10G
BLDPBD54595-100GEA
123
EUR
BLDPBD54595-100G
BLDPBD54595-10G
2-Chloro-1,4-dibromobenzene 98%
2-Chloro-1,4-dibromobenzene
Storage condition: Sealed in dry, Room Temperature
Formula:
C₆H₃Br₂Cl Storage Temperature: Ambient |
MDL Number:
MFCD00018094 CAS Number: 3460-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...