4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester 97%

BD553999-100MG BD553999-250MG
BLDPBD553999-100MGEA 55.7 EUR
BLDPBD553999-100MG BLDPBD553999-250MG
4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester 97%
4-Fluoro-2-hydroxybenzeneboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₂H₁₆BFO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16994248
CAS Number: 1038828-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR