tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenethylcarbamate 95%
|
|
BD556330-100MG
BD556330-250MG
BLDPBD556330-100MGEA
41
EUR
BLDPBD556330-100MG
BLDPBD556330-250MG
tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenethylcarbamate 95%
tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenethylcarbamate
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₉H₃₀BNO₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD22415459 CAS Number: 360792-43-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...