Motesanib diphosphate 98%

BD559247-1MG BD559247-10MG
BLDPBD559247-1MGEA 72.5 EUR
BLDPBD559247-1MG BLDPBD559247-10MG
Motesanib diphosphate 98%
Motesanib diphosphate

Storage condition: Inert atmosphere, Store in freezer, under -20°C

Formula: C₂₂H₂₉N₅O₉P₂
MW: 569.4 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12407403
CAS Number: 857876-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR